CAS RN 119-75-5
IUPAC Name:
2-nitro-N-phenylaniline
Synonyms:
2-nitrodiphenylamine
Molecular Weight (g/mol):
214.22
Molecular Formula:
C12H10N2O2
InChi Key:
RUKISNQKOIKZGT-UHFFFAOYSA-N
SMILES:
[O-][N+](=O)C1=CC=CC=C1NC1=CC=CC=C1
1
–
1
of
1
results
1
(2-Nitrophenyl)phenylamine, TRC
CAS: 119-75-5 Molecular Formula: C12 H10 N2 O2 Molecular Weight (g/mol): 214.22 Synonym: Benzenamine, 2-nitro-N-phenyl- (9CI, ACI),2-Nitro-N-phenylbenzenamine (ACI),Diphenylamine, 2-nitro- (8CI),2-Nitro-N-phenylaniline,2-Nitrodiphenylamine,C.I. 10335,N-(2-Nitrophenyl)aniline,N-Phenyl-2-nitroaniline,N-Phenyl-2-nitrobenzenamine,N-Phenyl-o-nitroaniline,NSC 105613,NSC 4754,NSC 629274,o-(Phenylamino)nitrobenzene,o-Nitro-N-phenylaniline,o-Nitrodiphenylamine,Sudan Yellow 1339 IUPAC Name: 2-nitro-N-phenylaniline SMILES: [O-][N+](=O)c1ccccc1Nc2ccccc2