accessibility menu, dialog, popup

UserName

CAS RN 119-91-5

NN

CAS RN 119-91-5

IUPAC Name: 2,2'-biquinoline
Synonyms: 2,2'-biquinoline
Molecular Weight (g/mol): 256.31
Molecular Formula: C18H12N2
InChi Key: WPTCSQBWLUUYDV-UHFFFAOYSA-N
SMILES: C1=CC2=CC=C(N=C2C=C1)C1=NC2=CC=CC=C2C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)