accessibility menu, dialog, popup

UserName

MgH2CH3CCH2CH3

CAS RN 1191-47-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 138.54
Molecular Formula: C8H18Mg
InChi Key: KJJBSBKRXUVBMX-UHFFFAOYSA-N
SMILES: [Mg++].CCC[CH2-].CCC[CH2-]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results