accessibility menu, dialog, popup

UserName

CAS RN 1192755-14-8

IUPAC Name: 2-(1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(1-benzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 244.1
Molecular Formula: C14H17BO3
InChi Key: DNZVVIZYTCIYNY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2OC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results