accessibility menu, dialog, popup

UserName

H3CSO

CAS RN 1193-82-4

IUPAC Name: methanesulfinylbenzene
Synonyms: methyl phenyl sulfoxide
Molecular Weight (g/mol): 140.2
Molecular Formula: C7H8OS
InChi Key: JXTGICXCHWMCPM-UHFFFAOYNA-N
SMILES: CS(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)