accessibility menu, dialog, popup

UserName

NaONHONNHN

CAS RN 1196-43-6

IUPAC Name: sodium 2,6-dioxo-2,3,6,9-tetrahydro-1H-purin-9-ide
Synonyms: sodium 2,6-dioxo-3,9-dihydro-1H-purin-9-ide
Molecular Weight (g/mol): 174.1
Molecular Formula: C5H3N4NaO2
InChi Key: HKDXLLDCXWUUKY-UHFFFAOYSA-M
SMILES: [Na+].O=C1NC2=C(N=C[N-]2)C(=O)N1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results