accessibility menu, dialog, popup

UserName

CAS RN 1198-30-7

NN

CAS RN 1198-30-7

IUPAC Name: isoquinoline-1-carbonitrile
Synonyms: isoquinoline-1-carbonitrile
Molecular Weight (g/mol): 154.17
Molecular Formula: C10H6N2
InChi Key: HJHXYSBRTVFEDD-UHFFFAOYSA-N
SMILES: N#CC1=C2C=CC=CC2=CC=N1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)