accessibility menu, dialog, popup

UserName

CAS RN 12007-99-7

BBBBBBCa

CAS RN 12007-99-7

IUPAC Name: calcium pentacyclo[3.1.0.0¹,³.0²,⁴.0⁴,⁶]hexaborane-1,4-diuide
Synonyms: calcium hexaboride
Molecular Weight (g/mol): 104.94
Molecular Formula: B6Ca
InChi Key: AWVIDOFQNLOVRC-UHFFFAOYSA-N
SMILES: [Ca++].B12B3[B-]11B4B1[B-]234
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)
  • (9)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)