accessibility menu, dialog, popup

UserName

BBBBSiBB

CAS RN 12008-29-6

IUPAC Name: 1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
Synonyms: 1-sila-2,3,4,5,6,7-hexaborapentacyclo[3.2.0.0¹,⁴.0²,⁷.0³,⁶]heptane
Molecular Weight (g/mol): 92.95
Molecular Formula: B6Si
InChi Key: ZRBFEDMQRDRUDG-UHFFFAOYSA-N
SMILES: B12B3B4B5B(B14)[Si]235

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results