CAS RN 12037-58-0
CAS RN 12037-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tungsten oxide, 99.99% (metals basis)
CAS: 12037-58-0 Molecular Formula: O2W Molecular Weight (g/mol): 215.84 MDL Number: MFCD00049709 InChI Key: DZKDPOPGYFUOGI-UHFFFAOYSA-N IUPAC Name: dioxotungsten SMILES: O=[W]=O