CAS RN 1205548-03-3
CAS RN 1205548-03-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4'-Dimethylamino 7,8-Dihydroxyflavone Dimethyl Ether, TRC
CAS: 1205548-03-3 Molecular Formula: C19H19NO4 Molecular Weight (g/mol): 325.36 Synonym: 2-[4-(Dimethylamino)phenyl]-7,8-dimethoxy-4H-1-benzopyran-4-one SMILES: Br.COc1ccc2C(=O)C=C(Oc2c1OC)c3ccc(cc3)N(C)C