CAS RN 120686-17-1
CAS RN 120686-17-1
IUPAC Name:
(2R)-4-(tert-butoxy)-4-oxo-1-phenylbutan-2-aminium
Synonyms:
(2R)-4-(tert-butoxy)-4-oxo-1-phenylbutan-2-aminium
Molecular Weight (g/mol):
236.33
Molecular Formula:
C14H22NO2
InChi Key:
ZIJHIHDFXCNFAA-GFCCVEGCSA-O
SMILES:
CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1
1
–
1
of
1
results
1
tert-Butyl (3S)-3-amino-4-phenylbutanoate, 97%
CAS: 120686-17-1 Molecular Formula: C14H22NO2 Molecular Weight (g/mol): 236.33 MDL Number: MFCD00798308 InChI Key: ZIJHIHDFXCNFAA-GFCCVEGCSA-O PubChem CID: 1501868 IUPAC Name: tert-butyl (3S)-3-amino-4-phenylbutanoate SMILES: CC(C)(C)OC(=O)C[C@H]([NH3+])CC1=CC=CC=C1