accessibility menu, dialog, popup

UserName

CAS RN 12070-10-9

CV

CAS RN 12070-10-9

IUPAC Name: methane vanadium
Synonyms: vanadium carbide
Molecular Weight (g/mol): 66.99
Molecular Formula: CH4V
InChi Key: GORXZVFEOLUTMI-UHFFFAOYSA-N
SMILES: C.[V]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results