accessibility menu, dialog, popup

UserName

CAS RN 12070-13-2

CWW

CAS RN 12070-13-2

IUPAC Name: methyliumylidyneditungstenene
Synonyms: methyliumylidyneditungstenene
Molecular Weight (g/mol): 379.69
Molecular Formula: CW2
InChi Key: FHLFYDOXSZQEPO-UHFFFAOYSA-N
SMILES: [W-]=[W]#[C+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results