CAS RN 1207995-62-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Nitroso-N,N-di(3,5,5-trimethylhexyl)amine (>85%), TRC
CAS: 1207995-62-7 Molecular Formula: C18 H38 N2 O Molecular Weight (g/mol): 298.5071 Synonym: 3,5,5-Trimethyl-N-nitroso-N-(3,5,5-trimethylhexyl)-1-hexanamine,1-Hexanamine, 3,5,5-trimethyl-N-nitroso-N-(3,5,5-trimethylhexyl)-,N-Nitrosodiisononylamine IUPAC Name: N,N-bis(3,5,5-trimethylhexyl)nitrous amide SMILES: CC(CCN(CCC(C)CC(C)(C)C)N=O)CC(C)(C)C