CAS RN 120801-75-4
IUPAC Name:
trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Synonyms:
trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Molecular Weight (g/mol):
250.26
Molecular Formula:
C5H9F3O4SSi
InChi Key:
XHVSCKNABCCCAC-UHFFFAOYSA-N
SMILES:
C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O
1
–
1
of
1
results
1
Trimethylsilyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 94%
CAS: 120801-75-4 Molecular Formula: C5H9F3O4SSi Molecular Weight (g/mol): 250.26 MDL Number: MFCD02093343 InChI Key: XHVSCKNABCCCAC-UHFFFAOYSA-N PubChem CID: 2778022 IUPAC Name: trimethylsilyl 2,2-difluoro-2-fluorosulfonylacetate SMILES: C[Si](C)(C)OC(=O)C(F)(F)S(F)(=O)=O