accessibility menu, dialog, popup

UserName

OOH3CH3CRh

CAS RN 12082-47-2

IUPAC Name: bis(ethene) 4-oxopent-2-en-2-olate rhodium
Synonyms: bis(ethylene) 4-oxopent-2-en-2-olate rhodium
Molecular Weight (g/mol): 258.12
Molecular Formula: C9H15O2Rh
InChi Key: AFQSOHSPTULSFS-UHFFFAOYSA-M
SMILES: [Rh].C=C.C=C.CC([O-])=CC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)