CAS RN 12093-05-9
IUPAC Name:
cycloocta-1,3,5,7-tetraene tris(methanidylidyneoxidanium) iron
Synonyms:
1,3,5,7-cyclooctatetraene tris(carbon monoxide) iron
Molecular Weight (g/mol):
244.03
Molecular Formula:
C11H8FeO3
InChi Key:
XAOCYVWRKJTXOL-UHFFFAOYSA-N
SMILES:
[Fe].[C-]#[O+].[C-]#[O+].[C-]#[O+].C1=CC=CC=CC=C1
1
–
1
of
1
results
1
(Cyclooctatetraene)iron tricarbonyl, 98%
CAS: 12093-05-9 Molecular Formula: C11H8FeO3 Molecular Weight (g/mol): 244.027 MDL Number: MFCD00001755 InChI Key: XAOCYVWRKJTXOL-JGZYGLCTSA-N Synonym: cyclooctatetraene iron tricarbonyl,tricarbonyl cyclooctatetraene iron,cyclooctatetraenetricarbonyliron,tricarbonyl cyclooctatetraene iron ii,1,3,5,7-cyclooctatetraeneiron tricarbonyl,tris carbon monoxide cyclooctatetraene iron PubChem CID: 11436476 IUPAC Name: carbon monoxide;cyclooctatetraene;iron SMILES: [C-]#[O+].[C-]#[O+].[C-]#[O+].C1=CC=CC=CC=C1.[Fe]