CAS RN 1212078-33-5
CAS RN 1212078-33-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1S)-1-[4-(Propan-2-yloxy)phenyl]ethan-1-amine, TRC
CAS: 1212078-33-5 Molecular Formula: C11H17NO Molecular Weight (g/mol): 179.25 IUPAC Name: (1S)-1-(4-propan-2-yloxyphenyl)ethanamine SMILES: CC(C)Oc1ccc(cc1)[C@H](C)N