accessibility menu, dialog, popup

UserName

MoOOOHHH

CAS RN 12125-77-8

IUPAC Name: 1,1,1-tris(hydroxymethyl)-1-molybdahexacyclo[3.3.0.0¹,³.0¹,⁴.0¹,⁶.0²,⁸]octan-1-yl
Synonyms: 1,1,1-tris(hydroxymethyl)-1-molybdahexacyclo[3.3.0.0¹,³.0¹,⁴.0¹,⁶.0²,⁸]octan-1-yl
Molecular Weight (g/mol): 275.15
Molecular Formula: C10H11MoO3
InChi Key: ZWJSTNVCXCHPHF-UHFFFAOYNA-N
SMILES:
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results