accessibility menu, dialog, popup

UserName

CH3OOCOCOCOCr

CAS RN 12125-87-0

IUPAC Name: tris(methanidylidyneoxidanium) methyl benzoate chromium
Synonyms: tris(carbon monoxide) methyl benzoate chromium
Molecular Weight (g/mol): 272.18
Molecular Formula: C11H8CrO5
InChi Key: GEALMZJLYGNHRA-UHFFFAOYSA-N
SMILES: [Cr].[C-]#[O+].[C-]#[O+].[C-]#[O+].COC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results