accessibility menu, dialog, popup

UserName

Organo-Transition Metal Compounds

Organo-transition metal compounds possess direct bonds between carbon atoms and transition metal atoms. Available in various chemical compositions, quantities, and purities, they can be used as homogeneous catalysts and as intermediates in organic synthesis.
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Showing products from the Organo-Transition Metal Compounds category. View All Search Results

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)