CAS RN 121357-04-8
IUPAC Name:
2-(2-methyl-1,3-thiazol-4-yl)ethan-1-ol
Synonyms:
2-(2-methyl-1,3-thiazol-4-yl)ethanol
Molecular Weight (g/mol):
143.2
Molecular Formula:
C6H9NOS
InChi Key:
REKVZAFNJAMAQL-UHFFFAOYSA-N
SMILES:
CC1=NC(CCO)=CS1
1
–
1
of
1
results
1
2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Thermo Scientific™
CAS: 121357-04-8 Molecular Formula: C6H9NOS Molecular Weight (g/mol): 143.204 InChI Key: REKVZAFNJAMAQL-UHFFFAOYSA-N Synonym: 2-2-methyl-1,3-thiazol-4-yl ethanol,2-2-methylthiazol-4-yl ethanol,2-2-methyl-1,3-thiazol-4-yl ethan-1-ol,4-thiazoleethanol,2-methyl,2-2-methyl-4-thiazolyl ethanol,acmc-20duqt,2-methyl-4-thiazoleethanol,2-methylthiazole-4-ethanol,4-hydroxyethyl-2-methyl-1,3-thiazole PubChem CID: 19876739 IUPAC Name: 2-(2-methyl-1,3-thiazol-4-yl)ethanol SMILES: CC1=NC(=CS1)CCO