accessibility menu, dialog, popup

UserName

CAS RN 12137-20-1

CAS RN 12137-20-1

IUPAC Name: oxotitanium
Synonyms: titanium(II) oxide
Molecular Weight (g/mol): 63.87
Molecular Formula: OTi
InChi Key: OGIDPMRJRNCKJF-UHFFFAOYSA-N
SMILES: O=[Ti]
Quantity 
  • (3)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)