accessibility menu, dialog, popup

UserName

H3CH3CCH3ONNBr

CAS RN 121487-13-6

IUPAC Name: 5-bromo-2-(tert-butoxy)pyrimidine
Synonyms: 5-bromo-2-(tert-butoxy)pyrimidine
Molecular Weight (g/mol): 231.09
Molecular Formula: C8H11BrN2O
InChi Key: UPKQXRBCDUBUTI-UHFFFAOYSA-N
SMILES: CC(C)(C)OC1=NC=C(Br)C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results