accessibility menu, dialog, popup

UserName

CAS RN 1215321-14-4

IUPAC Name: 2-chloro-3-(difluoromethyl)quinoline
Synonyms: 2-chloro-3-(difluoromethyl)quinoline
Molecular Weight (g/mol): 213.61
Molecular Formula: C10H6ClF2N
InChi Key: HDIMNLCODJNJNI-UHFFFAOYSA-N
SMILES: FC(F)C1=CC2=CC=CC=C2N=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results