accessibility menu, dialog, popup

UserName

CAS RN 1215721-40-6

NNHNNOClHCl

CAS RN 1215721-40-6

IUPAC Name: 2-[(3-chlorophenyl)methoxy]-6-(piperazin-1-yl)pyrazine hydrochloride
Synonyms: 2-[(3-chlorophenyl)methoxy]-6-(piperazin-1-yl)pyrazine hydrochloride
Molecular Weight (g/mol): 341.24
Molecular Formula: C15H18Cl2N4O
InChi Key: NMUNRTCTDLORDR-UHFFFAOYSA-N
SMILES: Cl.ClC1=CC=CC(COC2=NC(=CN=C2)N2CCNCC2)=C1
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results