CAS RN 121654-84-0
CAS RN 121654-84-0
IUPAC Name:
thiane-4-carbonyl chloride
Synonyms:
thiane-4-carbonyl chloride
Molecular Weight (g/mol):
164.65
Molecular Formula:
C6H9ClOS
InChi Key:
RJRIIGFPFQOTNF-UHFFFAOYSA-N
SMILES:
ClC(=O)C1CCSCC1
No Refinements
1
–
0
of
0
results
1
Tetrahydrothiopyran-4-carbonyl chloride, 95%, Thermo Scientific™
CAS: 121654-84-0 Molecular Formula: C6H9ClOS Molecular Weight (g/mol): 164.647 MDL Number: MFCD09879922 InChI Key: RJRIIGFPFQOTNF-UHFFFAOYSA-N Synonym: tetrahydrothiopyran-4-carbonyl chloride,tetrahydro-thiopyran-4-carbonyl chloride,tetrahydro-2h-thiopyran-4-carbonyl chloride,2h-thiopyran-4-carbonylchloride, tetrahydro,acmc-20mplw PubChem CID: 14547368 IUPAC Name: thiane-4-carbonyl chloride SMILES: C1CSCCC1C(=O)Cl