CAS RN 121917-57-5
CAS RN 121917-57-5
IUPAC Name:
(1S,9R)-1-methyl-16-azatetracyclo[7.6.1.0²,⁷.0¹⁰,¹⁵]hexadeca-2,4,6,10,12,14-hexaene; (2E)-but-2-enedioic acid
Synonyms:
dizocilpine; fumaric acid
Molecular Weight (g/mol):
337.38
Molecular Formula:
C20H19NO4
InChi Key:
QLTXKCWMEZIHBJ-CXGSIYSVSA-N
SMILES:
OC(=O)\C=C\C(O)=O.C[C@]12N[C@H](CC3=CC=CC=C13)C1=CC=CC=C21
1
–
2
of
2
results
1
Tocris Bioscience™ (-)-MK 801 maleate
CAS: 121917-57-5 Molecular Formula: C20H19NO4
2
(-)-Dizocilpine maleate, MedChemExpress
MedChemExpress (-)-Dizocilpine maleate ((-)-MK-801 maleate) is a less active (-)-enantiomer of Dizocilpine. (-)-Dizocilpine maleate is a selective and non-competitive N-methyl-D-aspartate (NMDA) receptor antagonist with a Ki of 211.7 nM. (-)-Dizocilpine maleate has antidepressant effects.