accessibility menu, dialog, popup

UserName

ONOClO

CAS RN 122-04-3

IUPAC Name: 4-nitrobenzoyl chloride
Synonyms: benzoyl chloride, 4-nitro-
Molecular Weight (g/mol): 185.56
Molecular Formula: C7H4ClNO3
InChi Key: SKDHHIUENRGTHK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)