accessibility menu, dialog, popup

UserName

HOONNOHO

CAS RN 122-05-4

IUPAC Name: pyrazine-2,5-dicarboxylic acid
Synonyms: pyrazine-2,5-dicarboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: GMIOYJQLNFNGPR-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=C(C=N1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results