accessibility menu, dialog, popup

UserName

NNBrN

CAS RN 1221791-93-0

IUPAC Name: 8-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Synonyms: 8-bromoimidazo[1,2-a]pyridine-6-carbonitrile
Molecular Weight (g/mol): 222.05
Molecular Formula: C8H4BrN3
InChi Key: DWOXNHVFPVGULV-UHFFFAOYSA-N
SMILES: BrC1=CC(=CN2C=CN=C12)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results