accessibility menu, dialog, popup

UserName

CH3NNHOFFF

CAS RN 122431-37-2

IUPAC Name: 2-methyl-5-(trifluoromethyl)-2,3-dihydro-1H-pyrazol-3-one
Synonyms: 2-methyl-5-(trifluoromethyl)-1H-pyrazol-3-one
Molecular Weight (g/mol): 166.1
Molecular Formula: C5H5F3N2O
InChi Key: WQRHIGNAKDJJKN-UHFFFAOYSA-N
SMILES: CN1NC(=CC1=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)