CAS RN 1227675-51-5
CAS RN 1227675-51-5
IUPAC Name:
5-chloro-10-methyl-10-azatetracyclo[9.8.0.0²,⁷.0¹⁴,¹⁹]nonadeca-2,4,6,14,16,18-hexaen-4-ol hydrobromide
Synonyms:
5-chloro-10-methyl-10-azatetracyclo[9.8.0.0²,⁷.0¹⁴,¹⁹]nonadeca-2,4,6,14,16,18-hexaen-4-ol hydrobromide
Molecular Weight (g/mol):
394.74
Molecular Formula:
C19H21BrClNO
InChi Key:
GAUWIDFICGEZKR-UHFFFAOYNA-N
SMILES:
Br.CN1CCC2=CC(Cl)=C(O)C=C2C2C1CCC1=CC=CC=C21
1
–
1
of
1
results
1
Promotions Available
Tocris Bioscience™ SCH 39166 hydrobromide
Tocris Bioscience™ SCH 39166 hydrobromide
CAS: 1227675-51-5 Molecular Formula: C19H21BrClNO