accessibility menu, dialog, popup

UserName

RhClRhCl

CAS RN 12279-09-3

IUPAC Name: tetrakis(cyclooctene) dirhodium dichloride
Synonyms: chlorobis(cyclooctene)rhodium dimer
Molecular Weight (g/mol): 717.51
Molecular Formula: C32H56Cl2Rh2
InChi Key: ZFCBAJWXKUDJSW-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Rh].[Rh].C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results