accessibility menu, dialog, popup

UserName

CAS RN 123-17-1

H3CCH3CH3OHCH3CH3

CAS RN 123-17-1

IUPAC Name: 2,6,8-trimethylnonan-4-ol
Synonyms: 4-nonanol, 2,6,8-trimethyl-
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: LFEHSRSSAGQWNI-UHFFFAOYNA-N
SMILES: CC(C)CC(C)CC(O)CC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results