accessibility menu, dialog, popup

UserName

CAS RN 123333-84-6

IUPAC Name: lithium(1+) bis(propan-2-yl)azanide oxolane
Synonyms: lithium(1+) THF diisopropylazanide
Molecular Weight (g/mol): 179.23
Molecular Formula: C10H22LiNO
InChi Key: YTJXGDYAEOTOCG-UHFFFAOYSA-N
SMILES: [Li+].C1CCOC1.CC(C)[N-]C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results