accessibility menu, dialog, popup

UserName

CAS RN 123409-01-8

SOOOSOOOSOOONaNaNa

CAS RN 123409-01-8

IUPAC Name: naphthalene-1,3,6-trisulfonic acid hydrate trisodium
Synonyms: naphthalene-1,3,6-trisulfonic acid hydrate trisodium
Molecular Weight (g/mol): 455.33
Molecular Formula: C10H10Na3O10S3
InChi Key: BMCNZRJPDRPEDI-UHFFFAOYSA-N
SMILES: O.[Na].[Na].[Na].OS(=O)(=O)C1=CC=C2C(=C1)C=C(C=C2S(O)(=O)=O)S(O)(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results