CAS RN 1235-21-8
CAS RN 1235-21-8
IUPAC Name:
(2-oxopropyl)triphenylphosphanium chloride
Synonyms:
(2-oxopropyl)triphenylphosphanium chloride
Molecular Weight (g/mol):
354.81
Molecular Formula:
C21H20ClOP
InChi Key:
XAMZZEBAJZJERT-UHFFFAOYSA-M
SMILES:
[Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Acetonyltriphenylphosphonium chloride, 99%
CAS: 1235-21-8 Molecular Formula: C21H20ClOP Molecular Weight (g/mol): 354.81 MDL Number: MFCD00011813 InChI Key: XAMZZEBAJZJERT-UHFFFAOYSA-M Synonym: acetonyltriphenylphosphonium chloride,2-oxopropyl triphenylphosphonium chloride,2-oxopropyltriphenylphosphonium chloride,phosphonium, acetonyltriphenyl-, chloride,acetonyl triphenylphosphonium chloride,phosphonium, 2-oxopropyl triphenyl-, chloride,acetonyltriphenylphosphoniumchloride,2-oxopropyl triphenylphosphanium chloride,1-triphenylphosphino acetone, chloride,acmc-1c01z PubChem CID: 196994 IUPAC Name: 2-oxopropyl(triphenyl)phosphanium;chloride SMILES: [Cl-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1