accessibility menu, dialog, popup

UserName

CH3CCCCCH3CH3CCH3CH3H3COCH3OCH3OTi

CAS RN 123927-75-3

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 276.2
Molecular Formula: C13H24O3Ti
InChi Key: GQQYDESAAOEXPK-UHFFFAOYSA-N
SMILES: [Ti+4].C[O-].C[O-].C[O-].C[C]1[C](C)[C](C)[C](C)[C]1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results