accessibility menu, dialog, popup

UserName

CAS RN 1240261-82-8

FSFFFFFOOH(E/Z)

CAS RN 1240261-82-8

IUPAC Name: 3-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]prop-2-enoic acid
Synonyms: 3-[3-fluoro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 292.2
Molecular Formula: C9H6F6O2S
InChi Key: KNRGBEHCOVNLOW-UHFFFAOYSA-N
SMILES: OC(=O)C=CC1=CC(F)=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results