accessibility menu, dialog, popup

UserName

NOON

CAS RN 1249094-81-2

IUPAC Name: 4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]benzonitrile
Synonyms: 4-[(2,5-dioxopyrrol-1-yl)methyl]benzonitrile
Molecular Weight (g/mol): 212.21
Molecular Formula: C12H8N2O2
InChi Key: DFJVGKSXXPQRLG-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1CC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results