accessibility menu, dialog, popup

UserName

CAS RN 124959-28-0

IUPAC Name: 2-chloro-4-(furan-2-yl)pyrimidine
Synonyms: 2-chloro-4-(furan-2-yl)pyrimidine
Molecular Weight (g/mol): 180.59
Molecular Formula: C8H5ClN2O
InChi Key: BLFCXYJTVFTNRX-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(=N1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results