accessibility menu, dialog, popup

UserName

NOOONNH3CCH3CH3CH3FBFFF

CAS RN 125700-73-4

IUPAC Name: [(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium; tetrafluoroboranuide
Synonyms: [(dimethylamino)({3,5-dioxo-4-azatricyclo[5.2.1.0²,⁶]dec-8-en-4-yl}oxy)methylidene]dimethylazanium tetrafluoroborate
Molecular Weight (g/mol): 365.14
Molecular Formula: C14H20BF4N3O3
InChi Key: DMTWWFCCBWWXSA-UHFFFAOYNA-N
SMILES: F[B-](F)(F)F.CN(C)C(ON1C(=O)C2C3CC(C=C3)C2C1=O)=[N+](C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results