CAS RN 1259198-59-8
CAS RN 1259198-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2Beta)-Methyl Megestrol Acetate, TRC
CAS: 1259198-59-8 Molecular Formula: C25H34O4 Molecular Weight (g/mol): 398.54 Synonym: (2β)-17-(Acetyloxy)-2,6-dimethylpregna-4,6-diene-3,20-dione,Megestrol Acetate EP Impurity G IUPAC Name: [(8R,9S,10R,13S,14S,17R)-17-acetyl-2,6,10,13-tetramethyl-3-oxo-2,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate SMILES: CC1C[C@@]2([C@H]3CC[C@]4([C@H]([C@@H]3C=C(C2=CC1=O)C)CC[C@@]4(C(=O)C)OC(=O)C)C)C