accessibility menu, dialog, popup

UserName

CAS RN 126-91-0

CH3CH3(R)H3COHCH2

CAS RN 126-91-0

IUPAC Name: (3R)-3,7-dimethylocta-1,6-dien-3-ol
Synonyms: (-)-linalool P-linalool D-allo-ocimenol
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: CDOSHBSSFJOMGT-JTQLQIEISA-N
SMILES: CC(C)=CCC[C@@](C)(O)C=C
Boiling Point 
  • (2)
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)