CAS RN 1262776-24-8
CAS RN 1262776-24-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Clothianidin-d3 (N-methyl-d3), TRC
CAS: 1262776-24-8 Molecular Formula: C6 2H3 H5 Cl N5 O2 S Molecular Weight (g/mol): 252.7 Synonym: 1-[(2-Chloro-1,3-thiazol-5-yl)methyl]-2-nitro-3-(trideuteriomethyl)guanidine IUPAC Name: 1-[(2-chloro-1,3-thiazol-5-yl)methyl]-2-nitro-3-(trideuteriomethyl)guanidine SMILES: [2H]C([2H])([2H])N\C(=N/[N+](=O)[O-])\NCc1cnc(Cl)s1
2
Clothianidin-d3 (N-methyl-d3), CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.