CAS RN 1263278-79-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dichloro Trazodone Monohydrochloride, TRC
CAS: 1263278-79-0 Molecular Formula: C19H21Cl2N5O . HCl Molecular Weight (g/mol): 442.8 Synonym: 2-[3-[4-(3,4-Dichlorophenyl)-1-piperazinyl]propyl]-1,2,4-triazolo[4,3-a]pyridin-3(2H)-one Monohydrochloride; IUPAC Name: 2-[3-[4-(3,4-dichlorophenyl)piperazin-1-yl]propyl]-[1,2,4]triazolo[4,3-a]pyridin-3-one;hydrochloride SMILES: Cl.Clc1ccc(cc1Cl)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2