accessibility menu, dialog, popup

UserName

CAS RN 127142-66-9

IUPAC Name: 1-chloro-4-isothiocyanato-2-nitrobenzene
Synonyms: 1-chloro-4-isothiocyanato-2-nitrobenzene
Molecular Weight (g/mol): 214.62
Molecular Formula: C7H3ClN2O2S
InChi Key: ZXGZBHIDSJXKLE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1Cl)N=C=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results