accessibility menu, dialog, popup

UserName

CAS RN 12741-05-8

ClClClY

CAS RN 12741-05-8

IUPAC Name: yttrium(3+) hydrate trichloride
Synonyms: yttrium(3+) hydrate trichloride
Molecular Weight (g/mol): 213.27
Molecular Formula: Cl3H2OY
InChi Key: BWOAAAYNOWWMHL-UHFFFAOYSA-K
SMILES: O.[Cl-].[Cl-].[Cl-].[Y+3]
Grade 
  • (6)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)